This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other A830080D01RIK SIRNA Assays ›
Gene Symbol:
A830080D01Rik
Gene Name:
RIKEN cDNA A830080D01 gene
Gene Aliases:
2310067E24Rik, Gm1146
Chromosome Location:
Chr.X: 159532641 - 159593081 on Build GRCm38
UniGene ID:
Mm.296254
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS222164
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: A830080D01Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033472.2 NP_001028644.2 9 2036
GenBank mRNA BC075674.1 - 9 1998

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001127568, XM_006256938, XM_006256936, XM_006256934, XM_006256933, and others.

Panther Classification:

Molecular Function -

nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline