siRNA Details
| Gene Symbol: Tmtc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198967.5 | NP_945318.2 | 14, 15 | 2220 | ||
| GenBank mRNA | AK046396.1 | - | 13, 14 | 2107 | ||
| AK137545.1 | - | 14, 15 | 2192 | |||
| AK162239.1 | - | 15, 16 | 1960 | |||
| BC023818.1 | - | 3, 4 | 381 | |||
| BC145513.1 | - | 12, 13 | 1552 | |||

Tmtc1
Genomic Map