siRNA Details
| Gene Symbol: Ercc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015769.2 | NP_056584.2 | 8, 9 | 2016 | ||
| GenBank mRNA | AF189285.1 | - | 8, 9 | 2016 | ||
| BC026792.1 | - | 8, 9 | 1807 | 4911382 | ||
| CD352377.1 | - | 1, 2 | 276 | |||
| CX210668.1 | - | 1, 2 | 170 | |||

Ercc4
Genomic Map