This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other EXOC7 SIRNA Assays ›
Gene Symbol:
Exoc7
Gene Name:
exocyst complex component 7
Gene Aliases:
Exo70, sec70
Chromosome Location:
Chr.11: 116287998 - 116306749 on Build GRCm38
UniGene ID:
Mm.22530
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS225366
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Exoc7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162872.1 NP_001156344.1 13 1488
NM_016857.2 NP_058553.2 15 1620
GenBank mRNA AF014461.1 - 15 1555
AK087601.1 - 13 1476
AK140450.1 - 15 1594
AK167769.1 - 13 1471
AK169928.1 - 13 1475
AK173094.1 - 13 6107
BC028927.1 - 13 1464 4208803
BC053710.1 - 13 1475 30056570

Back To Top

More Information


Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

transport
exocytosis
protein transport
regulation of entry of bacterium into host cell
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-79bds:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline