siRNA Details
| Gene Symbol: Cpb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019775.3 | NP_062749.2 | 11 | 1216 | ||
| GenBank mRNA | AB021968.1 | - | 11 | 1164 | ||
| AF164524.1 | - | 11 | 1231 | |||
| AF186188.1 | - | 11 | 1167 | |||
| AK004045.1 | - | 11 | 1189 | |||
| AK015095.1 | - | 955 | ||||
| BC089577.1 | - | 11 | 1188 | 6766672 | ||
| CO807338.1 | - | 3 | 250 | |||

Cpb2
Genomic Map