siRNA Details
| Gene Symbol: Sdhaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025333.4 | NP_079609.2 | 4 | 584 | ||
| GenBank mRNA | AK013454.1 | - | 4 | 489 | ||
| AK075594.1 | - | 4 | 489 | |||
| AK160175.1 | - | 2 | 217 | |||
| AK160957.1 | - | 4 | 491 | |||
| AK167228.1 | - | 4 | 520 | |||
| BC029630.1 | - | 3 | 303 | |||
| BC038924.1 | - | 3 | 552 | |||
| BC048913.1 | - | 4 | 317 | |||
| BC085277.1 | - | 4 | 486 | 30441476 | ||

Sdhaf2
Genomic Map