siRNA Details
| Gene Symbol: Pspc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025682.3 | NP_079958.3 | 6 | 1293 | ||
| GenBank mRNA | AK011272.1 | - | 6 | 1290 | ||
| AK017689.1 | - | 6 | 1294 | |||
| AK150823.1 | - | 6 | 1294 | |||
| AK160722.1 | - | 6 | 1294 | |||
| BC026772.1 | - | 6 | 1272 | 3711631 | ||

Pspc1
Genomic Map