siRNA Details
| Gene Symbol: Pkp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026163.2 | NP_080439.1 | 6 | 1410 | ||
| GenBank mRNA | AK004650.1 | - | 6 | 1411 | ||
| AK004731.2 | - | 6 | 1410 | |||
| AK005020.1 | - | 6 | 1442 | |||
| AK049481.1 | - | 6 | 1411 | |||
| AK167660.1 | - | 6 | 1412 | |||
| BC068279.1 | - | 4 | 690 | |||
| BC138252.1 | - | 6 | 1410 | 8861271 | ||
| BC138253.1 | - | 6 | 1410 | 8861272 | ||

Pkp2
Genomic Map