This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FRMD8 SIRNA Assays ›
Gene Symbol:
Frmd8
Gene Name:
FERM domain containing 8
Gene Aliases:
1200004M23Rik, 2310035N23Rik, 4931429L16Rik, AU018809
Chromosome Location:
Chr.19: 5850974 - 5875270 on Build GRCm38
UniGene ID:
Mm.202092
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS228363
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price Contact Us
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Frmd8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026169.4 NP_080445.1 10 1367
GenBank mRNA AK004603.2 - 10 1390
AK011659.1 - 5 707
AK018201.1 - 1 154
AK049224.1 - 10 1382
AK049233.1 - 10 1382
AK083902.1 - 10 1369
AK088639.1 - 10 1379
AK148434.1 - 1379
AK168669.1 - 10 1378
BC004765.1 - 10 1435 3156148

Back To Top

More Information


Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-sp9vq:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline