This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other COL6A4 SIRNA Assays ›
Gene Symbol:
Col6a4
Gene Name:
collagen, type VI, alpha 4
Gene Aliases:
1110001D15Rik, AI413310, AU023415, Dvwa, EG235580, Vwa6
Chromosome Location:
Chr.9: 105990318 - 106096691 on Build GRCm38
UniGene ID:
Mm.28854
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS229177
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Col6a4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026763.2 NP_081039.2 8 3434
GenBank mRNA AB370265.1 - 8 3526
AM231153.1 - 2 814

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
protein heterotrimerization
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline