This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other MFSD11 SIRNA Assays ›
Gene Symbol:
Mfsd11
Gene Name:
major facilitator superfamily domain containing 11
Gene Aliases:
2600014M03Rik
Chromosome Location:
Chr.11: 116852283 - 116875660 on Build GRCm38
UniGene ID:
Mm.348004
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS229957
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mfsd11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178620.3 NP_848735.1 5 464
GenBank mRNA AF015191.1 - 5 430
AK031384.1 - 5 659
AK032043.1 - 5 660
BC016209.1 - 5 423
BC079583.1 - 5 463
BC088988.1 - 5 464 6418757
BC100547.1 - 6 596
BC137970.1 - 5 451 8860989
BC137971.1 - 5 452 8860990
BC145068.1 - 5 403

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-7p2bx:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline