siRNA Details
| Gene Symbol: Scrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027268.2 | NP_081544.1 | 3, 4 | 484 | ||
| GenBank mRNA | AK012765.1 | - | 3, 4 | 512 | ||
| AK032573.1 | - | 3, 4 | 511 | |||
| AK050481.1 | - | 3, 4 | 502 | |||
| AK082953.1 | - | 3, 4 | 484 | |||
| AK089968.1 | - | 3, 4 | 474 | |||
| AK129084.1 | - | 3, 4 | 471 | |||
| AK145904.1 | - | 3, 4 | 486 | |||
| BC023889.1 | - | 3, 4 | 471 | 5252046 | ||
| EF410099.1 | - | 435 | ||||

Scrn1
Genomic Map