siRNA Details
| Gene Symbol: Ipo5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023579.4 | NP_076068.1 | 10 | 1124 | ||
| GenBank mRNA | AK049422.1 | - | 10 | 1096 | ||
| AK053681.1 | - | 10 | 1089 | |||
| AK079134.1 | - | 10 | 1094 | |||
| AK167303.1 | - | 10 | 1086 | |||
| AK169664.1 | - | 10 | 1092 | |||
| BC020012.1 | - | 4 | 396 | |||
| BC051433.1 | - | 3 | 169 | |||
| BC052392.1 | - | 10 | 1067 | 5719784 | ||
| BC054814.1 | - | 10 | 1297 | 6842146 | ||

Ipo5
Genomic Map