This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other INTS10 SIRNA Assays ›
Gene Symbol:
Ints10
Gene Name:
integrator complex subunit 10
Gene Aliases:
4921521J11Rik, AI462004
Chromosome Location:
Chr.8: 68793928 - 68829410 on Build GRCm38
UniGene ID:
Mm.252421
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS230488
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ints10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293791.1 NP_001280720.1 3, 4 383
NM_001293792.1 NP_001280721.1 3, 4 383
NM_027590.4 NP_081866.1 3, 4 383
GenBank mRNA AK014939.1 - 3, 4 359
AK034498.1 - 3, 4 299
AK040574.1 - 3, 4 384
AK047093.1 - 3, 4 378
AK147743.1 - 3, 4 357

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

snRNA processing
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline