siRNA Details
| Gene Symbol: Osbpl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081434.1 | NP_001074903.1 | 20 | 2215 | ||
| GenBank mRNA | AK017095.1 | - | 2 | 236 | ||
| AK154677.1 | - | 20 | 2253 | |||
| AK171585.1 | - | 20 | 2244 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Osbpl7| Gene Symbol: Osbpl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081434.1 | NP_001074903.1 | 20 | 2215 | ||
| GenBank mRNA | AK017095.1 | - | 2 | 236 | ||
| AK154677.1 | - | 20 | 2253 | |||
| AK171585.1 | - | 20 | 2244 | |||