siRNA Details
| Gene Symbol: Myo1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181072.3 | NP_851417.2 | 18 | 2209 | ||
| GenBank mRNA | AF426465.1 | - | 1 | 63 | ||
| AK029145.1 | - | 18 | 2211 | |||
| BC040786.1 | - | 19 | 2265 | |||
| BC051391.1 | - | 18 | 2191 | 5053090 | ||
| BG086565.2 | - | 1 | 29 | |||
| BY761514.1 | - | 5 | 408 | |||

Myo1e
Genomic Map