This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other KIF1BP SIRNA Assays ›
Gene Symbol:
Kif1bp
Gene Name:
KIF1 binding protein
Gene Aliases:
0710007C18Rik, 2510003E04Rik, KBP, mKIAA1279
Chromosome Location:
Chr.10: 62558470 - 62578464 on Build GRCm38
UniGene ID:
Mm.258955
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS231507
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kif1bp
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028197.2 NP_082473.2 7 1299
GenBank mRNA AK129320.1 - 7 1284
AK141324.1 - 7 1308
AK149629.1 - 7 1236
AK158730.1 - 8 1033
BC038828.1 - 3 608
BC046485.1 - 7 1270 5350638
BC055307.1 - 7 1253 6493547
BC090974.1 - 7 1253 6774777

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitochondrial transport
multicellular organism development
nervous system development
cell differentiation
kinesin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-fg2nk:80/100.66.79.138:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline