This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CEP55 SIRNA Assays ›
Gene Symbol:
Cep55
Gene Name:
centrosomal protein 55
Gene Aliases:
1200008O12Rik, 2700032M20Rik
Chromosome Location:
Chr.19: 38054981 - 38074425 on Build GRCm38
UniGene ID:
Mm.9916
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS232310
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cep55
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164362.1 NP_001157834.1 8, 9 1508
NM_028293.1 NP_082569.1 7, 8 1132
NM_028760.2 NP_083036.2 8, 9 1484
GenBank mRNA AK004655.1 - 8, 9 1510
AK028216.1 - 7, 8 1118
AK088548.1 - 8, 9 1191
BC026966.1 - 5, 6 840
BC031396.1 - 8, 9 1437 4950530

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitotic cytokinesis
cell separation after cytokinesis
cell cycle
mitotic nuclear division
establishment of protein localization
cell division
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-ftbch:80/100.66.79.106:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0