This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 4930451G09RIK SIRNA Assays ›
Gene Symbol:
4930451G09Rik
Gene Name:
RIKEN cDNA 4930451G09 gene
Gene Aliases:
-
Chromosome Location:
Chr.16: 4964289 - 4978962 on Build GRCm38
UniGene ID:
Mm.374835
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS232824
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 4930451G09Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271586.1 NP_001258515.1 8, 9 1372
GenBank mRNA AK015442.1 - 8, 9 1368
AK078576.1 - 5, 6 742
AK154225.1 - 8, 9 1292

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001014115, XM_006245807, XM_006245810, XM_006245809, BC083674.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-5gjhx:80/100.66.77.87:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0