siRNA Details
| Gene Symbol: Vmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029478.3 | NP_083754.2 | 6 | 597 | ||
| GenBank mRNA | AB047550.1 | - | 6 | 609 | ||
| AB076609.1 | - | 5 | 488 | |||
| AK016312.1 | - | 6 | 614 | |||
| AK043091.1 | - | 6 | 598 | |||
| AK049559.1 | - | 6 | 590 | |||
| AK050796.1 | - | 5 | 466 | |||
| AK077443.1 | - | 6 | 597 | |||
| AK159468.1 | - | 6 | 612 | |||
| AK167173.1 | - | 6 | 605 | |||
| AK167202.1 | - | 6 | 605 | |||
| AK189413.1 | - | 2 | 177 | |||
| AK216058.1 | - | 2 | 176 | |||
| AX775963.1 | - | 6 | 567 | |||
| BC004013.1 | - | 6 | 591 | 3493738 | ||

Vmp1
Genomic Map