This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TMEM200A SIRNA Assays ›
Gene Symbol:
Tmem200a
Gene Name:
transmembrane protein 200A
Gene Aliases:
C030003D03Rik
Chromosome Location:
Chr.10: 25991182 - 26079408 on Build GRCm38
UniGene ID:
Mm.266260
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS233918
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem200a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029881.3 NP_084157.2 2 2390
GenBank mRNA AK032569.1 - 2 1990
AK048673.1 - 3 1785
AK141454.1 - 2362
AK165025.1 - 2 1558
AK173299.1 - 2 2390
BC141266.1 - 2 1615 9055878
BC141267.1 - 2 1615 9055879

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-v4fgr:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline