This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ESCO1 SIRNA Assays ›
Gene Symbol:
Esco1
Gene Name:
establishment of cohesion 1 homolog 1 (S. cerevisiae)
Gene Aliases:
A930014I12Rik
Chromosome Location:
Chr.18: 10566507 - 10610685 on Build GRCm38
UniGene ID:
Mm.210996
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS234090
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price Contact Us
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Esco1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081222.1 NP_001074691.1 10 3333
GenBank mRNA AK040576.1 - 10 1161
AK049589.1 - 9 1050
AK145333.1 - 10 3334
AK145870.1 - 9 981
AK173297.1 - 8 2711
BC008220.1 - 9 1205
BC076613.1 - 10 3063
BC151069.1 - 10 3289 9087982
BC151077.1 - 10 3289 9087990
BC172153.1 - 11 3411

Back To Top

More Information


Panther Classification:

Molecular Function -

acetyltransferase

Gene Ontology Categories:

Function(s) Process(es)

regulation of DNA replication
cell cycle
sister chromatid cohesion
post-translational protein acetylation
molecular_function
dihydrolipoamide branched chain acyltransferase activity
sterol O-acyltransferase activity
N-acetyltransferase activity
O-acyltransferase activity
palmitoleoyl [acyl-carrier-protein]-dependent acyltransferase activity
carnitine O-acyltransferase activity
acetyltransferase activity
C-acyltransferase activity
palmitoyltransferase activity
N-acyltransferase activity
acylglycerol O-acyltransferase activity
serine O-acyltransferase activity
O-acetyltransferase activity
O-octanoyltransferase activity
octanoyltransferase activity
O-palmitoyltransferase activity
S-acyltransferase activity
S-acetyltransferase activity
S-malonyltransferase activity
malonyltransferase activity
C-palmitoyltransferase activity
transferase activity
transferase activity, transferring acyl groups
succinyltransferase activity
N-succinyltransferase activity
O-succinyltransferase activity
S-succinyltransferase activity
sinapoyltransferase activity
O-sinapoyltransferase activity
peptidyl-lysine N6-myristoyltransferase activity
peptidyl-lysine N6-palmitoyltransferase activity
benzoyl acetate-CoA thiolase activity
3-hydroxybutyryl-CoA thiolase activity
3-ketopimelyl-CoA thiolase activity
N-palmitoyltransferase activity
myristoyltransferase activity
acyl-CoA N-acyltransferase activity
protein-cysteine S-myristoyltransferase activity
protein-cysteine S-acyltransferase activity
dihydrolipoamide S-acyltransferase activity
glucosaminyl-phosphotidylinositol O-acyltransferase activity
ergosterol O-acyltransferase activity
lanosterol O-acyltransferase activity
naphthyl-2-oxomethyl-succinyl-CoA succinyl transferase activity
2,4,4-trimethyl-3-oxopentanoyl-CoA 2-C-propanoyl transferase activity
2-methylhexanoyl-CoA C-acetyltransferase activity
butyryl-CoA 2-C-propionyltransferase activity
2,6-dimethyl-5-methylene-3-oxo-heptanoyl-CoA C-acetyltransferase activity
L-2-aminoadipate N-acetyltransferase activity
UDP-3-O-[3-hydroxymyristoyl] glucosamine N-acyltransferase activity
keto acid formate lyase activity
Ras palmitoyltransferase activity
metal ion binding
azetidine-2-carboxylic acid acetyltransferase activity
peptidyl-lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
acetyl-CoA:L-lysine N6-acetyltransferase

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0