This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HOOK1 SIRNA Assays ›
Gene Symbol:
Hook1
Gene Name:
hook homolog 1 (Drosophila)
Gene Aliases:
A930033L17Rik, azh
Chromosome Location:
Chr.4: 95967187 - 96024277 on Build GRCm38
UniGene ID:
Mm.49994
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS234130
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hook1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030014.2 NP_084290.1 11 1142
GenBank mRNA AF487912.1 - 11 1224
AK028714.1 - 11 1161
AK043870.1 - 11 1149
AK049897.1 - 11 1143
BC061688.1 - 11 1142 5364426

Back To Top

More Information


Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

transport
endosome organization
lysosome organization
multicellular organism development
spermatogenesis
spermatid development
endosome to lysosome transport
protein transport
cell differentiation
early endosome to late endosome transport
actin binding
protein binding
microtubule binding
identical protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-qj4g5:80/100.66.75.65:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0