This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HEMGN SIRNA Assays ›
Gene Symbol:
Hemgn
Gene Name:
hemogen
Gene Aliases:
4921524M03Rik, AI317176, EDAG, Hgn, mNDR
Chromosome Location:
Chr.4: 46393989 - 46413504 on Build GRCm38
UniGene ID:
Mm.25793
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS235034
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price Contact Us
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hemgn
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053149.2 NP_444379.1 3, 4 1573
GenBank mRNA AF269248.1 - 3, 4 1573
AK076587.1 - 4, 5 1558
AK077692.1 - 3, 4 1416
AK156536.1 - 3, 4 1501
AK171890.1 - 3, 4 1492
AK171906.1 - 3, 4 1628
AK172421.1 - 3, 4 1493
AY699986.1 - 3, 4 1415
BC050798.1 - 4, 5 1552 6743819

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
cell differentiation
regulation of osteoblast differentiation
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0