siRNA Details
| Gene Symbol: Smyd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160127.1 | NP_001153599.1 | 10 | 1503 | ||
| NM_009762.2 | NP_033892.2 | 9 | 1464 | |||
| GenBank mRNA | AK076266.1 | - | 9 | 1429 | ||
| AK142153.1 | - | 9 | 1432 | |||
| AK142252.1 | - | 10 | 1481 | |||
| BC076601.1 | - | 10 | 1481 | 30609878 | ||
| U76371.1 | - | 1608 | ||||
| U76373.2 | - | 10 | 1475 | |||
| U76374.2 | - | 9 | 1436 | |||

Smyd1
Genomic Map