siRNA Details
| Gene Symbol: Pex11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162387.1 | NP_001155859.1 | 4 | 729 | ||
| NM_001162388.1 | NP_001155860.1 | 4 | 493 | |||
| NM_011069.3 | NP_035199.3 | 4 | 729 | |||
| GenBank mRNA | AF093671.1 | - | 4 | 474 | ||
| AK005236.1 | - | 4 | 481 | |||
| AK011709.1 | - | 4 | 527 | |||
| AK082279.1 | - | 4 | 488 | |||
| AK082778.1 | - | 4 | 450 | |||
| BC013812.1 | - | 4 | 478 | 3483892 | ||
| BC054414.1 | - | 4 | 477 | 3257217 | ||
| BQ443231.1 | - | 4 | 539 | |||

Pex11b
Genomic Map