siRNA Details
| Gene Symbol: LOC101055749 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX636598.1 | - | 1 | 168 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101055749| Gene Symbol: LOC101055749 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX636598.1 | - | 1 | 168 | ||