This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FBXW19 SIRNA Assays ›
Gene Symbol:
Fbxw19
Gene Name:
F-box and WD-40 domain protein 19
Gene Aliases:
E330023O18
Chromosome Location:
Chr.9: 109478507 - 109495863 on Build GRCm38
UniGene ID:
Mm.133921
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS239313
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price Contact Us
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fbxw19
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177703.3 NP_808371.1 10 1361
GenBank mRNA AK087808.1 - 10 1362
AK162088.1 - 142

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-ftbch:80/100.66.79.106:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0