siRNA Details
| Gene Symbol: Dmtf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110327.1 | NP_001103797.1 | 13 | 1518 | ||
| NM_011806.3 | NP_035936.3 | 14 | 1728 | |||
| GenBank mRNA | AK029147.1 | - | 14 | 1670 | ||
| AK039563.1 | - | 13 | 1486 | |||
| AK046201.1 | - | 13 | 1589 | |||
| AK078015.1 | - | 13 | 1421 | |||
| AK160595.1 | - | 13 | 1491 | |||
| BC045141.1 | - | 13 | 1442 | 6307203 | ||
| BC060109.1 | - | 14 | 1650 | |||

Dmtf1
Genomic Map