siRNA Details
| Gene Symbol: Slc26a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178743.3 | NP_848858.2 | 13 | 1415 | ||
| GenBank mRNA | AF345196.1 | - | 13 | 1415 | ||
| AK052202.1 | - | 8 | 1502 | |||
| AK089182.1 | - | 13 | 1395 | |||
| BC038604.1 | - | 13 | 1408 | |||
| BC095937.1 | - | 9 | 1381 | |||
| BC132493.1 | - | 8 | 1283 | 40130770 | ||
| BC137897.1 | - | 8 | 1283 | 8860916 | ||

Slc26a11
Genomic Map