siRNA Details
| Gene Symbol: Irf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252382.1 | NP_001239311.1 | 9 | 1562 | ||
| NM_012057.4 | NP_036187.1 | 9 | 1510 | |||
| GenBank mRNA | AF028725.1 | - | 9 | 1402 | ||
| AK150598.1 | - | 9 | 1429 | |||
| AK151019.1 | - | 9 | 1427 | |||
| AK152042.1 | - | 9 | 1429 | |||
| AK156220.1 | - | 9 | 1562 | |||
| EU401974.1 | - | 6 | 1061 | |||

Irf5
Genomic Map