siRNA Details
| Gene Symbol: BC051142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001177.2 | NP_001001177.2 | 19, 20 | 1035 | ||
| NM_001163855.1 | NP_001157327.1 | 22, 23 | 784 | |||
| GenBank mRNA | AK077077.1 | - | 19, 20 | 1022 | ||
| AK133031.1 | - | 18, 19 | 868 | |||
| BC051142.1 | - | 22, 23 | 784 | 6740688 | ||

BC051142
Genomic Map