This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RHOX10 SIRNA Assays ›
Gene Symbol:
Rhox10
Gene Name:
reproductive homeobox 10
Gene Aliases:
-
Chromosome Location:
Chr.X: 38066475 - 38071688 on Build GRCm38
UniGene ID:
Mm.244139
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS243539
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price Contact Us
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rhox10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024850.2 NP_001020021.1 1 140
GenBank mRNA AK146090.1 - 1 122
BC100366.1 - 1 84 30524902
BC132474.1 - 1 71 40130751
BC132500.1 - 1 71 40130777
DQ058647.1 - 1 140

Back To Top

More Information


Panther Classification:

Molecular Function -

homeodomain transcription factor

Gene Ontology Categories:

Function(s)

biological_process

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rr9bx:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0