This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PSMD10 SIRNA Assays ›
Gene Symbol:
Psmd10
Gene Name:
proteasome (prosome, macropain) 26S subunit, non-ATPase, 10
Gene Aliases:
AW554874
Chromosome Location:
Chr.X: 140948425 - 140956711 on Build GRCm38
UniGene ID:
Mm.17640
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS244073
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price Contact Us
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Psmd10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164177.1 NP_001157649.1 5 632
NM_016883.4 NP_058579.2 5 711
GenBank mRNA AB022022.1 - 5 667
AK018233.1 - 5 688
AK136400.1 - 5 690
BC026931.1 - 5 678 3987112
BC056196.1 - 5 696 30019985

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
apoptotic process
cytoplasmic sequestering of NF-kappaB
positive regulation of cell growth
positive regulation of protein ubiquitination
negative regulation of NF-kappaB transcription factor activity
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
negative regulation of apoptotic process
negative regulation of MAPK cascade
negative regulation of DNA damage response, signal transduction by p53 class mediator
positive regulation of cyclin-dependent protein serine/threonine kinase activity
proteasome regulatory particle assembly
negative regulation of release of cytochrome c from mitochondria
transcription factor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ndfxr:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline