siRNA Details
| Gene Symbol: Rbm18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159635.1 | NP_001153107.1 | 4, 5 | 468 | ||
| NM_026434.4 | NP_080710.1 | 4, 5 | 558 | |||
| GenBank mRNA | AK008110.1 | - | 4, 5 | 414 | ||
| AK021243.1 | - | 4, 5 | 439 | |||
| AK035348.1 | - | 4, 5 | 466 | |||
| AK036252.1 | - | 4, 5 | 439 | |||
| AK135109.1 | - | 3, 4 | 339 | |||
| BC005540.1 | - | 4, 5 | 452 | 3590164 | ||
| CB183820.1 | - | 4, 5 | 629 | |||

Rbm18
Genomic Map