This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CPN2 SIRNA Assays ›
Gene Symbol:
Cpn2
Gene Name:
carboxypeptidase N, polypeptide 2
Gene Aliases:
1300018K11Rik
Chromosome Location:
Chr.16: 30256378 - 30267614 on Build GRCm38
UniGene ID:
Mm.290614
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS245776
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cpn2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027904.3 NP_082180.2 2 601
GenBank mRNA AK005049.1 - 2 603
BC025836.1 - 2 568
BC081550.1 - 1 120
BC138287.1 - 2 688 8861306
BC138288.1 - 2 688 8861307

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107085, XM_006248485, BC160859.

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline