This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SYCE2 SIRNA Assays ›
Gene Symbol:
Syce2
Gene Name:
synaptonemal complex central element protein 2
Gene Aliases:
1700013H19Rik, AA407907, Cesc1
Chromosome Location:
Chr.8: 84872111 - 84887446 on Build GRCm38
UniGene ID:
Mm.229128
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS245798
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price Contact Us
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Syce2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168244.1 NP_001161716.1 2, 3 726
NM_001168246.1 NP_001161718.1 3, 4 425
NM_027954.3 NP_082230.2 3, 4 407
GenBank mRNA AK005954.1 - 3, 4 408
AK082833.1 - 2, 3 293
AK167332.1 - 2, 3 725
BC094628.1 - 3, 4 302 3374002
BC132534.1 - 3, 4 357 40130811
BY246600.1 - 3, 4 340

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001191557, XM_006255291.

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
synaptonemal complex assembly
cell division
meiotic cell cycle
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-gm6kg:80/100.66.79.106:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0