siRNA Details
| Gene Symbol: Lingo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013758.2 | NP_001013780.2 | 2 | 1152 | ||
| GenBank mRNA | BC072620.1 | - | 2 | 1143 | 6815693 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lingo3| Gene Symbol: Lingo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013758.2 | NP_001013780.2 | 2 | 1152 | ||
| GenBank mRNA | BC072620.1 | - | 2 | 1143 | 6815693 | |