This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other OLFR1305 SIRNA Assays ›
Gene Symbol:
Olfr1305
Gene Name:
olfactory receptor 1305
Gene Aliases:
MOR245-8
Chromosome Location:
Chr.2: 111872915 - 111873853 on Build GRCm38
UniGene ID:
Mm.377480
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS248457
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olfr1305
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146401.2 NP_666513.2 1 32
GenBank mRNA BC106975.1 - 1 25
BC106976.1 - 1 25
BC114640.1 - 1 31

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001000607.

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception
detection of chemical stimulus involved in sensory perception of smell
transmembrane signaling receptor activity
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline