siRNA Details
| Gene Symbol: Prex2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033636.4 | NP_001028808.1 | 3 | 361 | ||
| NM_029525.1 | NP_083801.1 | 37 | 4878 | |||
| GenBank mRNA | AK136059.1 | - | 3 | 362 | ||
| AM109952.1 | - | 37 | 4879 | |||
| BC139253.1 | - | 3 | 321 | |||
| BC139255.1 | - | 3 | 321 | |||

Prex2
Genomic Map