This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other C1S1 SIRNA Assays ›
Gene Symbol:
C1s1
Gene Name:
complement component 1, s subcomponent 1
Gene Aliases:
AA959438, AI255193, AI327365, C1s, C1sa
Chromosome Location:
Chr.6: 124530344 - 124542390 on Build GRCm38
UniGene ID:
Mm.219527
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS249893
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C1s1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001097617.1 NP_001091086.1 12 1758
NM_144938.2 NP_659187.2 12 1795
GenBank mRNA AF459019.1 - 12 1722
AK087522.1 - 12 1727
AK172453.1 - 12 1723
BC018319.1 - 12 1687 4196654
BC022123.1 - 12 1748 5097206
BC111880.1 - 1551 40059749

Back To Top

More Information


Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

immune system process
proteolysis
complement activation
complement activation, classical pathway
innate immune response
serine-type endopeptidase activity
calcium ion binding
peptidase activity
serine-type peptidase activity
hydrolase activity
identical protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-fbxsq:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline