siRNA Details
| Gene Symbol: C1s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001097617.1 | NP_001091086.1 | 12 | 2101 | ||
| NM_144938.2 | NP_659187.2 | 12 | 2138 | |||
| GenBank mRNA | AF459019.1 | - | 12 | 2065 | ||
| AK087522.1 | - | 12 | 2070 | |||
| AK172453.1 | - | 12 | 2066 | |||
| BC018319.1 | - | 12 | 2030 | 4196654 | ||
| BC022123.1 | - | 12 | 2091 | 5097206 | ||
| BC111880.1 | - | 1894 | 40059749 | |||

C1s1
Genomic Map