siRNA Details
| Gene Symbol: 4930519G04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026263.2 | NP_080539.1 | 7 | 640 | ||
| GenBank mRNA | AK005973.1 | - | 3 | 245 | ||
| AK015843.1 | - | 7 | 642 | |||
| AK015857.1 | - | 7 | 691 | |||
| AK161377.1 | - | 5 | 1541 | |||

4930519G04Rik
Genomic Map