This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 4930519G04RIK SIRNA Assays ›
Gene Symbol:
4930519G04Rik
Gene Name:
RIKEN cDNA 4930519G04 gene
Gene Aliases:
1700014B12Rik
Chromosome Location:
Chr.5: 114852411 - 114883880 on Build GRCm38
UniGene ID:
Mm.251409
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS250309
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 4930519G04Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026263.2 NP_080539.1 6 560
GenBank mRNA AK005973.1 - 2 165
AK015843.1 - 6 562
AK015857.1 - 6 611
AK161377.1 - 4 1461

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-qgwn9:80/100.66.78.68:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0