This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CAAP1 SIRNA Assays ›
Gene Symbol:
Caap1
Gene Name:
caspase activity and apoptosis inhibitor 1
Gene Aliases:
5830433M19Rik, AI536248, CAAP
Chromosome Location:
Chr.4: 94460277 - 94557076 on Build GRCm38
UniGene ID:
Mm.35170
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS250345
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Caap1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026368.2 NP_080644.2 4, 5 687
GenBank mRNA AK076487.1 - 4, 5 642
AK140382.1 - 4, 5 688
AK150311.1 - 5, 6 597
AK160297.1 - 4, 5 690
AK160572.1 - 4, 5 688
AK171141.1 - 4, 5 688
BC020067.1 - 4, 5 675 3991510

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001034154, XM_008763894, XM_006238400, BC088436, BC105829.

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-7svs5:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline