siRNA Details
| Gene Symbol: Cenpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011942.1 | - | 2, 3 | 532 | ||
| AK150539.1 | - | 2, 3 | 650 | |||
| AK151163.1 | - | 2, 3 | 650 | |||
| AK154611.1 | - | 2, 3 | 601 | |||
| BC020064.1 | - | 2, 3 | 556 | |||
| BC060191.1 | - | 2, 3 | 525 | |||
| BC080823.1 | - | 2, 3 | 477 | |||
| BC116944.1 | - | 2, 3 | 575 | 40126263 | ||
| BC116970.1 | - | 2, 3 | 575 | 40126289 | ||

Cenpl
Genomic Map