This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 1700001O22RIK SIRNA Assays ›
Gene Symbol:
1700001O22Rik
Gene Name:
RIKEN cDNA 1700001O22 gene
Gene Aliases:
1700113K14Rik, 4930549G18Rik
Chromosome Location:
Chr.2: 30795563 - 30803724 on Build GRCm38
UniGene ID:
Mm.34166
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS250744
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 1700001O22Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198000.3 NP_932117.2 2 619
GenBank mRNA AK005618.1 - 2 130
AK007200.1 - 108
AK019754.1 - 2 96
AK076928.1 - 2 120
BC023044.1 - 2 589 5355351

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0