This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GM11744 SIRNA Assays ›
Gene Symbol:
Gm11744
Gene Name:
predicted gene 11744
Gene Aliases:
OTTMUSG00000003947, Prcd
Chromosome Location:
Chr.11: 116657108 - 116668389 on Build GRCm38
UniGene ID:
Mm.390349
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS270044
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm11744
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163318.1 NP_001156790.1 1 32
GenBank mRNA AK137302.1 - 463
DQ390337.1 - 1 32

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_006247840, XM_006247841.

Gene Ontology Categories:

Function(s) Process(es)

visual perception
biological_process
response to stimulus
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mlg7h:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline