This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other AKR1C14 SIRNA Assays ›
Gene Symbol:
Akr1c14
Gene Name:
aldo-keto reductase family 1, member C14
Gene Aliases:
9030611N15Rik, AW557061
Chromosome Location:
Chr.13: 4048999 - 4090560 on Build GRCm38
UniGene ID:
Mm.26838
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS272383
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price Contact Us
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Akr1c14
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134072.1 NP_598833.1 7 803
GenBank mRNA AK033537.1 - 7 821
AK136528.1 - 7 820
AY730283.1 - 7 765
BC013482.1 - 7 803 4240772
BC026628.1 - 7 802 4983237

Back To Top

More Information


Panther Classification:

Molecular Function -

oxidoreductase reductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

oxidation-reduction process
steroid dehydrogenase activity
androsterone dehydrogenase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-ftgjf:80/100.66.79.187:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline