This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PTTG1IP SIRNA Assays ›
Gene Symbol:
Pttg1ip
Gene Name:
pituitary tumor-transforming 1 interacting protein
Gene Aliases:
1810010L20Rik, AI314311, AU018448, C79540
Chromosome Location:
Chr.10: 77581767 - 77598732 on Build GRCm38
UniGene ID:
Mm.28853
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS272647
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pttg1ip
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145925.2 NP_666037.1 3, 4 330
GenBank mRNA AK035696.1 - 3, 4 345
AK075773.1 - 3, 4 303
AK088813.1 - 3, 4 367
AK089887.1 - 3, 4 312
AK098093.1 - 3, 4 344
AK151639.1 - 3, 4 312
AK152366.1 - 3, 4 312
AK159987.1 - 3, 4 312
AK167972.1 - 3, 4 342
AK168164.1 - 3, 4 342
AK170802.1 - 3, 4 312
AK170971.1 - 3, 4 345
AK170972.1 - 3, 4 312
AK171140.1 - 3, 4 311
AK191979.1 - 1, 2 30
BC023961.1 - 3, 4 385 5323397
BC025533.1 - 3, 4 354 5250502
BC029144.1 - 3, 4 326 4945974
BC032184.1 - 3, 4 349 5310191

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein import into nucleus
positive regulation of protein ubiquitination
negative regulation of DNA damage response, signal transduction by p53 class mediator
negative regulation of intrinsic apoptotic signaling pathway by p53 class mediator
positive regulation of cellular protein catabolic process
p53 binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0